Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP1B4 Rabbit anti-Human, Clone: 108, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP29015850UL
Description
ATP1B4 Monoclonal specifically detects ATP1B4 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
ATP1B4 | |
Monoclonal | |
Unconjugated | |
0.2 um filtered solution in PBS | |
Rabbit | |
Protein A purified | |
RUO | |
23439 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, ELISA, Immunoprecipitation | |
108 | |
Western Blot 1:500-1:1000, ELISA 1:25000-1:50000, Immunoprecipitation 0.2-1 ul/mg of lysate | |
ATPase, (Na+)/K+ transporting, beta 4 polypeptide, ATPase, Na+/K+ transporting, beta 4 polypeptide, K-ATPase beta-m subunit, K-ATPase subunit beta-m, protein ATP1B4, X, X/potassium-transporting ATPase subunit beta-m | |
Recombinant Human ATP1B4 protein | |
50 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction