Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP2A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101857
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101857 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101857 Supplier Invitrogen™ Supplier No. PA5101857
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ATP2A1.

ATP dependent calcium pumps are responsible, in part, for the maintenance of low cytoplasmic free calcium concentrations. The ATP pumps that reside in intracellular organelles are encoded by a family of structurally related enzymes, termed the sarcoplasmic or endoplasmic reticulum calcium (SERCA) ATPases. The SERCA1 gene is exclusively expressed in type II (fast) skeletal muscle. The SERCA2 gene is subject to tissue dependent processing which is responsible for the generation of SERCA2a muscle-specific form expressed in type I (slow) skeletal, cardiac and smooth muscle and the SERCA2b isoform expressed in all cell types. The SERCA3 gene is not as well characterized and is found in non-muscle cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP2A1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ATP2A1
Gene Accession No. O14983, Q64578, Q8R429
Gene Alias adult Ca2+ ATPase; ATP2A; ATP2A1; ATP2A3; ATPase sarcoplasmic/endoplasmic reticulum Ca2+ transporting 1; ATPase, Ca++ transporting, cardiac muscle, fast twitch 1; ATPase, Ca++ transporting, ubiquitous; Ca2+ ATPase; calcium pump 1; Calcium-transporting ATPase sarcoplasmic reticulum type, fast twitch skeletal muscle isoform; endoplasmic reticulum class 1/2 Ca(2+) ATPase; neonatal Ca2+ ATPase; sarcoplasmic/endoplasmic reticulum calcium ATPase 1; SERCA; SERCA ATPase; Serca1; SERCA1a; SR Ca(2+)-ATPase 1
Gene Symbols ATP2A1
Host Species Rabbit
Immunogen A synthesized peptide derived from human ATP2A1(Accession O14983), corresponding to amino acid residues A547-V597.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116601, 11937, 487
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.