Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP2C1 Monoclonal Antibody (4G12)
GREENER_CHOICE

Catalog No. PIMA515592
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515592 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515592 Supplier Invitrogen™ Supplier No. MA515592
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15592 targets ATP2C1 in indirect ELISA, IHC and WB applications and shows reactivity with Human and Non-human primate samples. The MA5-15592 immunogen is purified recombinant fragment of ATP2C1 expressed in E. Coli. MA5-15592 detects ATP2C1 which has a predicted molecular weight of approximately 100kDa.

ATP2C1, also known as secretory pathway Ca2+/Mn2+-ATPase (SPCA) 1, belongs to the family of P-type cation transport ATPases. This magnesium-dependent enzyme catalyzes the hydrolysis of ATP coupled with the transport of the calcium from the cytosol to the Golgi lumen. Defects in this gene cause Hailey-Hailey disease, an autosomal dominant disorder characterized by persistent blisters and erosions of the skin. Unlike the related protein ATP2C2, ATP2C1 is ubiquitously expressed and displays a lower maximal turnover rate for overall Ca2+-ATPase reaction and a higher apparent affinity for cytosolic Ca2+ activation of phosphorylation. Recent evidence suggests that ATP2C1 is a key regulator of insulin-like growth factor receptor (IGF1R) processing in tumor progression in basal breast cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP2C1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 4G12
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene Atp2c1
Gene Accession No. P98194
Gene Alias 1700121J11Rik; ATP2C1; ATP2C1A; ATPase 2C1; ATPase secretory pathway Ca2+ transporting 1; ATPase, Ca(2+)-sequestering; ATPase, Ca++ transporting, type 2C, member 1; ATPase, Ca++-sequestering; ATP-dependent Ca(2+) pump PMR1; AW061228; BCPM; calcium-transporting ATPase 2C1; calcium-transporting ATPase type 2C member 1; D930003G21Rik; HHD; hSPCA1; HUSSY 28; HUSSY-28; KIAA1347; Pmr1; PMR1L; rat; secretory pathway Ca(2+)-ATPase 1; secretory pathway Ca2+/Mn2+ ATPase 1; SPCA; SPCA1
Gene Symbols Atp2c1
Host Species Mouse
Immunogen Purified recombinant fragment of ATP2C1 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27032
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.