Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP2C1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521126
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521126 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521126 Supplier Invitrogen™ Supplier No. PA521126
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is mouse brain tissue lysate. PA5-21126 can be used with blocking peptide PEP-1240.

ATP2C1, also known as secretory pathway Ca2+/Mn2+-ATPase (SPCA) 1, belongs to the family of P-type cation transport ATPases. This magnesium-dependent enzyme catalyzes the hydrolysis of ATP coupled with the transport of the calcium from the cytosol to the Golgi lumen. Defects in this gene cause Hailey-Hailey disease, an autosomal dominant disorder characterized by persistent blisters and erosions of the skin. Unlike the related protein ATP2C2, ATP2C1 is ubiquitously expressed and displays a lower maximal turnover rate for overall Ca2+-ATPase reaction and a higher apparent affinity for cytosolic Ca2+ activation of phosphorylation. Recent evidence suggests that ATP2C1 is a key regulator of insulin-like growth factor receptor (IGF1R) processing in tumor progression in basal breast cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP2C1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Atp2c1
Gene Accession No. P98194, Q80XR2
Gene Alias 1700121J11Rik; ATP2C1; ATP2C1A; ATPase 2C1; ATPase secretory pathway Ca2+ transporting 1; ATPase, Ca(2+)-sequestering; ATPase, Ca++ transporting, type 2C, member 1; ATPase, Ca++-sequestering; ATP-dependent Ca(2+) pump PMR1; AW061228; BCPM; calcium-transporting ATPase 2C1; calcium-transporting ATPase type 2C member 1; D930003G21Rik; HHD; hSPCA1; HUSSY 28; HUSSY-28; KIAA1347; Pmr1; PMR1L; rat; secretory pathway Ca(2+)-ATPase 1; secretory pathway Ca2+/Mn2+ ATPase 1; SPCA; SPCA1
Gene Symbols Atp2c1
Host Species Rabbit
Immunogen A 19 amino acid peptide near the carboxy terminus of human ATP2C1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 235574, 27032
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.