Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP2C2 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176567MFV500
Description
ATP2C2 Polyclonal antibody specifically detects ATP2C2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
ATP2C2 | |
Polyclonal | |
Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
ATPase 2C2, ATPase, Ca++ transporting, type 2C, member 2, calcium-transporting ATPase type 2C member 2, DKFZp686H22230, KIAA0703, secretory pathway Ca(2+)-ATPase 2, secretory pathway calcium ATPase 2, SPCA2 | |
Antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human ATP2C2. The immunogen is located within the first 50 amino acids of ATP2C2. Amino Acid Squence: RQYQALEKDEEEALIDEQS | |
0.1 mL | |
Cancer, Cell Cycle and Replication | |
9914 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunocytochemistry | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction