Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP2C2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562577
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562577 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562577 Supplier Invitrogen™ Supplier No. PA562577
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LEKDEEEALI DEQSELKAIE KEKKVTALPP KEACKCQKED LARAFCVDLH TGLSEFSVTQ RRLAHGWNEF VADNSEPVWK KYLDQ Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 76%.

ATP2C2, also known as secretory pathway Ca2+/Mn2+-ATPase (SPCA) 2, belongs to the family of P-type cation transport ATPases. This magnesium-dependent enzyme catalyzes the hydrolysis of ATP coupled with the transport of the calcium from the cytosol to the Golgi lumen. Defects in the related gene ATP2C1 cause Hailey-Hailey disease, for which ATP2C2 does not compensate, suggesting that ATP2C2 plays other physiological roles. Unlike ATP2C1, ATP2C2 has a much more restricted expression pattern and displays a higher maximal turnover rate for overall Ca2+-ATPase reaction and a lower apparent affinity for cytosolic Ca2+ activation of phosphorylation. Overexpression of ATP2C2 in mammary tumors result a Ca2+ influx via the store-operated Ca2+ channel ORAI1 and independent of the STIM1 and STIM2 sensors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP2C2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ATP2C2
Gene Accession No. O75185
Gene Alias 1810010G06Rik; Atp2c2; ATPase 2C2; ATPase secretory pathway Ca2+ transporting 2; ATPase, Ca++ transporting, type 2C, member 2; calcium-transporting ATPase type 2C member 2; Kiaa0703; mKIAA0703; Secretory pathway Ca(2+)-ATPase 2; secretory pathway Ca2+-ATPase; secretory pathway calcium ATPase 2; Spca2
Gene Symbols ATP2C2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ATP2C2. Recombinant protein control fragment (Product #RP-95779).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9914
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.