Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP4A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558673
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558673 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558673 Supplier Thermo Scientific Supplier No. PA558673
Add to Cart
Add to Cart

ATP4A Polyclonal Antibody

ATP4A Polyclonal Antibody for IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP4A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ATP4A
Gene Accession No. P20648
Gene Alias ATP4A; ATP6A; ATPase H+/K+ transporting alpha subunit; ATPase, H+/K+ exchanging, alpha polypeptide; ATPase, H+/K+ transporting, alpha polypeptide; gastric H(+)/K(+) ATPase subunit alpha; gastric H,K-ATPase alpha subunit; gastric H,K-ATPase catalytic subunit; gastric H+/K+ ATPase alpha subunit; gastric hydrogen-potassium ATPase; H(+)-K(+)-ATPase alpha subunit; potassium-transporting ATPase alpha chain 1; Proton pump
Gene Symbols ATP4A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ATP4A. Recombinant protein control fragment (Product #RP-95785).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 495
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.