Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP4B Mouse anti-Human, Pig, Clone: 2G11, Abnova™

Catalog No. 89035501 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-501 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-501 Supplier Abnova Corporation Supplier No. MAB14103

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against pig ATP4B.

  • High specificity
  • High reactivity

Specifications

Antigen ATP4B
Applications ChIP Assay, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2G11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against pig ATP4B.
Dilution ChIP Flow Cytometry (1:50) Immunocytochemistry Immunofluorescence (1:2000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:2000) Immunohistochemistry (Frozen sections) (1:2000) Immunoprecipitation Western Blot
Formulation In ascites (0.05% sodium azide)
Gene Accession No. P18434
Gene Symbols ATP4B
Host Species Mouse
Immunogen Purified 34 kDa core peptide corresponding to deglycosylated hog gastric microsomes.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 397131
Target Species Human, Pig
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.