Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP4B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587635
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587635 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587635 Supplier Invitrogen™ Supplier No. PA587635
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LMQTVDPYTP DYQDQLRSPG VTLRPDVYGE KGLEIVYNVS DNRTWADLTQ TLHAFLAGYS PAAQEDSINC TSEQYFFQES FRAPNHTKFS CKFTADMLQN CSGLADPNFG FEEGKPCFII KMNRIVKFLP SNGSAPRVDC AFLDQPRELG QPLQVKYYPP NGTFSLHYFP YYGKKAQPHY SNPLVAAKLL NIPRNAEVAI VCKVMAEHVT FNNPHDPYEG KVEFKLKIEK; Positive Samples: LO2, SW620, NCI-H460, Mouse small intestine, Mouse liver, Mouse heart, Mouse pancreas, Rat stomach; Cellular Location: Cell membrane, Single-pass type II membrane protein.

The hydrogen/potassium ATPase, or gastric proton pump, belongs to a family of P-type cation-transporting ATPases. This family of ATPases shares a number of functional and structural similarities including the common feature of consisting of an alpha and beta subunit. Like the ubiquitous sodium/potassium ATPase, the hydrogen/potassium ATPase consists of a large transmembrane catalytic subunit, termed the alpha- subunit which contains sites for ATP binding and phosphorylation, and an associated smaller glycoprotein, termed the beta-subunit which may play a role in maintaining the structural and functional integrity of the complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP4B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ATP4B
Gene Accession No. P18598, P50992, P51164
Gene Alias (H+,K+)-ATPase beta-subunit; ATP4B; ATP6B; ATPase H+/K+ transporting beta subunit; ATPase H+/K+ transporting subunit beta; ATPase H+K+-transporting beta / (gastric HK-ATPase beta subunit) defined by SSR; ATPase, H+,K+-transporting, beta / (gastric H,K-ATPase beta subunit), defined by SSR; ATPase, H+/K+ exchanging, beta polypeptide; ATPase, H+/K+ transporting, beta polypeptide; ATPase, H+/K+ transporting, beta polypeptide, gastric specific; AV080843; gastric H(+)/K(+) ATPase subunit beta; gastric H+/K+ ATPase beta subunit; gastric H+/K+-ATPase beta subunit; gastric hydrogen-potassium ATPase, beta; gp60-90; H,K-ATPase-Beta; H.K-ATPase, beta subunit; H+,K+-ATPase; H+/K+ ATPase beta; H+/K+ ATPase beta subunit; H+/K+-ATPase beta; H+/K+-ATPase beta subunit; potassium-transporting ATPase beta chain; potassium-transporting ATPase subunit beta; Proton pump beta chain; S76401S1
Gene Symbols ATP4B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 62-291 of human ATP4B (NP_0006961).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11945, 24217, 496
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.