Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP5A Antibody (5G11), Novus Biologicals™
SDP

Catalog No. p-200022692 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297994 50 μL
NB297995 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB297994 Supplier Novus Biologicals Supplier No. NBP32649150UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ATP5A Monoclonal antibody specifically detects ATP5A in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry

Specifications

Antigen ATP5A
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 5G11
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit 1, ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit, isoform1, cardiac muscle, ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit, isoform2, non-cardiac muscle-like 2, ATP sythase (F1-ATPase) alpha subunit, ATP5AL2, ATP5AMOM2, ATPMATP synthase subunit alpha, mitochondrial, cardiac muscle, EC 3.6.3, EC 3.6.3.14, hATP1, mitochondrial ATP synthetase, oligomycin-resistant, OMR, ORMATP synthase alpha chain, mitochondrial
Host Species Rabbit
Immunogen A synthesized peptide derived from Human ATP5A [UniProt P25705]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 498
Target Species Human, Mouse
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.