Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP5A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583505
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583505 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583505 Supplier Invitrogen™ Supplier No. PA583505
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QRELIIGDRQ TGKTSIAIDT IINQKRFNDG SDEKKKLYCI YVAIGQKRST VAQLVKRLTD ADAMKYTIVV SATASDAAPL QYLAP Highest antigen sequence indentity to the following orthologs: Rat - 99%, Mouse - 99%.

This gene encodes a subunit of mitochondrial ATP synthase. Mitochondrial ATP synthase catalyzes ATP synthesis, using an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled with a stoichiometry of 3 alpha, 3 beta, and a single representative of the other 3. The proton channel consists of three main subunits (a, b, c). This gene encodes the alpha subunit of the catalytic core. Alternatively spliced transcript variants encoding the different isoforms have been identified. Pseudogenes of this gene are located on chromosomes 9, 2, and 16.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP5A1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ATP5F1A
Gene Accession No. P25705
Gene Alias AI035633; AL022851; AL023067; alpha subunit ATP synthase isoform precursor (EC 3.6.1.34); ATP synthase alpha chain, mitochondrial; ATP synthase alpha subunit; ATP synthase alpha subunit precursor; ATP synthase F1 subunit alpha; ATP synthase subunit alpha heart isoform, mitochondrial; ATP synthase subunit alpha liver isoform, mitochondrial; ATP synthase subunit alpha, mitochondrial; ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit 1; ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit 1, cardiac muscle; ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit, isoform 1; ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit, isoform 1, cardiac muscle; ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit, isoform 2, non-cardiac muscle-like 2; ATP sythase (F1-ATPase) alpha subunit; ATP5A; Atp5a1; ATP5A2; ATP5AL2; ATP5F1A; atp5fa1; ATPM; cardiac muscle ATP synthase H+ transporting mitochondrial F1 complex alpha subunit 1; COXPD22; D18Ertd206e; EGK_09658; epididymis secretory sperm binding protein Li 123m; hATP1; HEL-S-123m; iso; MC5DN4; mitochondrial ATP synthase alpha chain; mitochondrial ATP synthase alpha subunit; mitochondrial ATP synthetase, oligomycin-resistant; mitochondrial H+-ATP synthase alpha subunit; modifier of Min 2; Mom2; OMR; ORM; zgc:154103
Gene Symbols ATP5A1, ATP5F1A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ATP5A1. Recombinant protein control fragment (Product #RP-102896).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 498
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.