Learn More
ATP5B Mouse anti-Canine, Human, Rat, Clone: OTI5G2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF500851

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ATP synthase is extremely conserved through evolution and can be found in plants, fungi, bacteria, and animals. The ATP synthase enzyme is a transmembrane protein responsible for driving the reversible reaction from ADP+ phosphate to ATP. This reaction is accomplished by a flux of protons across the membrane as a result of electron transfer. The ATP synthase protein has two main sections; the F1 ATP-ase (soluble) and the F0 ATP-ase (membrane embedded). The F1 section consists of the alpha, beta, gamma, delta, and epsilon subunits. While the F0 consists of a, b, and c subunits.Specifications
ATP5B | |
Monoclonal | |
Unconjugated | |
ATP5B | |
ATP5B, ATPMB, ATPSB, HEL-S-271 | |
Mouse | |
Affinity Chromatography | |
RUO | |
171374, 403669, 506 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI5G2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P06576, P10719, P99504 | |
ATP5B | |
Full length human recombit protein of human ATP5B produced in HEK293T cell. | |
100 μg | |
Primary | |
Canine, Human, Rat | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.