Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP5B Mouse anti-Human, Clone: 3D5AB1, Abnova™

Catalog No. 89035403 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-403 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-403 Supplier Abnova Corporation Supplier No. MAB13998

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against human ATP5B.

  • High specificity
  • High reactivity

Specifications

Antigen ATP5B
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 3D5AB1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human ATP5B.
Dilution Flow Cytometry (3-5 ug x 106 cells) Immunocytochemistry (2 ug/mL) Immunofluorescence (2 ug/mL) Western Blot (0.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In HEPES (0.02% sodium azide).
Gene Accession No. P06576
Gene Alias ATPMB/ATPSB/MGC5231
Gene Symbols ATP5B
Host Species Mouse
Immunogen Human heart mitochondria.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 506
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.