Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP5B Rabbit anti-Human, Mouse, Rat, Unconjugated, Clone: AADH-1 , Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB19596
Description
ATP5B Monoclonal antibody specifically detects ATP5B in Human, Mouse, Rat samples. It is validated for IP, ICC, WB-Ce, IHC-P, IF.Specifications
ATP5B | |
Monoclonal | |
Unconjugated | |
ATP5B | |
Rabbit | |
100 μg | |
506 | |
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
IgG |
Immunoprecipitation, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence | |
AADH-1 | |
ATP synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide | |
ATPMB, ATPSB, MGC5231 | |
A synthetic peptide corresponding to human ATP5B | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction