Learn More
Invitrogen™ ATP5D Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597291
Description
Immunogen sequence: MLPAALLRRP GLGRLVRHAR AYAEAAAAPA AASGPNQMSF TFASPTQVFF NGANVRQVDV PTLTGAFGIL AAHVPTLQVL RPGLVVVHAE DGTTSKYFVS SGSIAVNADS SVQLLAEEAV TLDMLDLGAA KANLEKAQAE LVGTADEATR AEIQIRIEAN EALVKALE; Positive Samples: MCF7, Raji, HepG2, A-549, Mouse kidney, Mouse heart, Mouse liver, Rat liver, Rat heart; Cellular Location: Mitochondrion, Mitochondrion inner membrane.
This gene encodes a subunit of mitochondrial ATP synthase. Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled with a stoichiometry of 3 alpha, 3 beta, and a single representative of the other 3. The proton channel consists of three main subunits (a, b, c). This gene encodes the delta subunit of the catalytic core. Alternatively spliced transcript variants encoding the same isoform have been identified.
Specifications
ATP5D | |
Polyclonal | |
Unconjugated | |
ATP5F1D | |
0610008F14Rik; 1500000I11Rik; AA960090; AI876556; ATP synthase F1 subunit delta; ATP synthase subunit delta, mitochondrial; ATP synthase, H+ transporting, mitochondrial F1 complex, delta subunit; ATP5D; ATP5F1D; AU020773; C85518; F-ATPase delta subunit; mitochondrial ATP synthase complex delta-subunit precusor; mitochondrial ATP synthase, delta subunit | |
Rabbit | |
Affinity Chromatography | |
RUO | |
245965, 513, 66043 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.64 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P30049, P35434, Q9D3D9 | |
ATP5F1D | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-168 of human ATP5D (NP_0016781). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.