Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP5D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597484
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597484 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597484 Supplier Invitrogen™ Supplier No. PA597484
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATP5D Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes a subunit of mitochondrial ATP synthase. Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled with a stoichiometry of 3 alpha, 3 beta, and a single representative of the other 3. The proton channel consists of three main subunits (a, b, c). This gene encodes the delta subunit of the catalytic core. Alternatively spliced transcript variants encoding the same isoform have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP5D
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.48 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ATP5F1D
Gene Accession No. P30049, Q9D3D9
Gene Alias 0610008F14Rik; 1500000I11Rik; AA960090; AI876556; ATP synthase F1 subunit delta; ATP synthase subunit delta, mitochondrial; ATP synthase, H+ transporting, mitochondrial F1 complex, delta subunit; ATP5D; ATP5F1D; AU020773; C85518; F-ATPase delta subunit; mitochondrial ATP synthase complex delta-subunit precusor; mitochondrial ATP synthase, delta subunit
Gene Symbols ATP5F1D
Host Species Rabbit
Immunogen Recombinant Human ATP synthase subunit delta, mitochondrial protein (1-168AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 513, 66043
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.