Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ATP5E Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104424
Description
Antibody detects endogenous levels of total ATP5E.
The Drosophila gene for the ATP synthase epsilon subunit (ATP5E) is known as stunted or sun (Kidd et al. 2005). Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled with a stoichiometry of 3 alpha, 3 beta, and a single representative of the other 3. The proton channel consists of three main subunits (a, b, c).
Specifications
ATP5E | |
Polyclonal | |
Unconjugated | |
ATP5F1E | |
2410043G19Rik; ATP synthase epsilon chain; ATP synthase epsilon chain, mitochondrial; ATP synthase F1 subunit delta L homeolog; ATP synthase F1 subunit epsilon; ATP synthase subunit epsilon, mitochondrial; ATP synthase, epsilon subunit; ATP synthase, H transporting, mitochondrial F1 complex, epsilon subunit; ATP synthase, H+ transporting, mitochondrial F1 complex, delta subunit L homeolog; ATP synthase, H+ transporting, mitochondrial F1 complex, epsilon subunit; atp5d; atp5d.L; Atp5e; atp5f1d.L; ATP5F1E; ATPase subunit epsilon; ATPE; ATPsynepsilon; AV000645; BcDNA:RE19513; BcDNA:RH48911; CG9032; CG9032-PA; CG9032-PB; Dmel\CG9032; Dmel_CG9032; epsilon subunit of ATP synthetase; F(0)F(1)-ATPase; F0/F1 ATP synthase epsilon; H(+)-transporting two-sector ATPase; l(1)13Fe; l(1)XV; LOC617230 protein; MC5DN3; mitochondrial ATP synthase epsilon chain; mitochondrial ATPase; Protein stunted; RE19513p; RH48911p; stunted; Sun; sun-PA; sun-PB; XELAEV_18006241mg | |
Rabbit | |
Affinity Chromatography | |
RUO | |
245958, 514, 67126 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P29418, P56381, P56382 | |
ATP5F1E | |
A synthesized peptide derived from human ATP5E(Accession P56381), corresponding to amino acid residues Q7-E51. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction