Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP5G1/2/3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114397
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114397 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114397 Supplier Invitrogen™ Supplier No. PA5114397
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human HEPG2 whole cell, human A549 whole cell, human PC-3 whole cell, human HL-60 whole cell, human HEK293 whole cell, human U937 whole cell, human CACO-2 whole cell, rat intestine tissue, human Ovarian cancer tissue, human tonsil tissue, human glioma tissue, human lung cancer tissue, human placenta tissue, mouse intestine tissue. IHC: human intestinal tissue. Flow: HL-60 cell, PC-3 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

The ATP5MC1 gene is one of three human paralogs that encode membrane subunit c of the mitochondrial ATP synthase. It is mapped to 17q21.32. This gene encodes a subunit of mitochondrial ATP synthase. Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled with a stoichiometry of 3 alpha, 3 beta, and a single representative of the other 3. The proton channel seems to have nine subunits (a, b, c, d, e, f, g, F6 and 8). This gene is one of three genes that encode subunit c of the proton channel. Each of the three genes have distinct mitochondrial import sequences but encode the identical mature protein. Alternatively spliced transcript variants encoding the same protein have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP5G1/2/3
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene ATP5G2
Gene Accession No. P05496, P48201, P56383, P56384, Q06055, Q06646, Q9CR84
Gene Alias 1810041M08Rik; 6030447M23; ATP synthase c subunit; ATP synthase F(0) complex subunit C1, mitochondrial; ATP synthase F(0) complex subunit C2, mitochondrial; ATP synthase F(0) complex subunit C3, mitochondrial; ATP synthase H+ transporting mitochondrial F0 complex subunit c (subunit 9) isoform 1; ATP synthase lipid-binding protein; ATP synthase lipid-binding protein, mitochondrial; ATP synthase membrane subunit c locus 1; ATP synthase membrane subunit c locus 2; ATP synthase membrane subunit c locus 3; ATP synthase proteolipid P1; ATP synthase proteolipid P2; ATP synthase proteolipid P3; ATP synthase proton-transporting mitochondrial F(0) complex subunit C1; ATP synthase proton-transporting mitochondrial F(0) complex subunit C2; ATP synthase proton-transporting mitochondrial F(0) complex subunit C3; ATP synthase subunit 9; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit c (subunit 9) isoform 3; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit c (subunit 9), isoform 1; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit c (subunit 9), isoform 2; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit c (subunit 9), isoform 3; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit c, isoform 1; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit c, isoform 3; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit C1 (subunit 9); ATP synthase, H+ transporting, mitochondrial F0 complex, subunit C2 (subunit 9); ATP synthase, H+ transporting, mitochondrial F0 complex, subunit C3 (subunit 9); ATP synthase, H+ transporting, mitochondrial Fo complex subunit C1 (subunit 9); ATP synthase, H+ transporting, mitochondrial Fo complex subunit C2 (subunit 9); ATP synthase, H+ transporting, mitochondrial Fo complex subunit C3 (subunit 9); ATP synthase, H+ transporting, mitochondrial Fo complex, subunit C1 (subunit 9); ATP synthase, H+ transporting, mitochondrial Fo complex, subunit C2 (subunit 9); ATP synthase, H+ transporting, mitochondrial Fo complex, subunit C3 (subunit 9); ATP synthase, mitochondrial, C subunit-3; ATP5A; ATP5G; ATP5G1; ATP5G2; Atp5g3; ATP5MC1; Atp5mc2; ATP5MC3; ATPase protein 9; ATPase subunit 9; ATPase subunit c; mitochondrial ATP synthase, subunit 9, isoform 1; mitochondrial ATP synthase, subunit C (subunit 9), isoform 2; mitochondrial ATP synthase, subunit C, isoform 1; P3; PSEC0033
Gene Symbols ATP5G1, ATP5G2, ATP5G3
Host Species Rabbit
Immunogen E.coli-derived human ATP5F1/ATP5MC1 recombinant protein (Position: D62-L113).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11951, 171082, 228033, 516, 517, 518, 67942
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.