Learn More
Description
Specifications
Specifications
| Antigen | ATP5G2 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Alias | ATP synthase lipid-binding protein, mitochondrial, ATP synthase proteolipid P2, ATP synthase, H+ transporting, mitochondrial F0 complex, subunit c (subunit 9), ATP synthase, H+ transporting, mitochondrial F0 complex, subunit C2 (subunit 9), ATP synthase, H+ transporting, mitochondrial Fo complex, subunit C2 (subunit 9), ATPase protein 9, ATPase subunit c, isoform 2, mitochondrial ATP synthase, subunit C (subunit 9) |
| Gene Symbols | ATP5G2 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against a recombinant protein corresponding to the amino acids: LCLLCSGSSSPATAPHPLKMFACSKFVSTPSLVKSTSQLLSRPLSAVVLKRPEILTDESLSSLAVSCPLTSLVSSRSFQ |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
