Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP5G2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562413
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562413 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562413 Supplier Invitrogen™ Supplier No. PA562413
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LCLLCSGSSS PATAPHPLKM FACSKFVSTP SLVKSTSQLL SRPLSAVVLK RPEILTDESL SSLAVSCPLT SLVSSRSFQ Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 58%.

ATP5G2, also named as ATPase protein 9 and ATPase subunit c, belongs to the ATPase C chain family. Mitochondrial membrane ATP synthase (F1F0 ATP synthase or Complex V) produces ATP from ADP in the presence of a proton gradient across the membrane which is generated by electron transport complexes of the respiratory chain. ATP5G2 is the major protein stored in the storage bodies of animals or humans affected with ceroid lipofuscinosis (Batten disease).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP5G2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ATP5MC2
Gene Accession No. Q06055
Gene Alias 1810041M08Rik; ATP synthase c subunit; ATP synthase F(0) complex subunit C2, mitochondrial; ATP synthase lipid-binding protein; ATP synthase lipid-binding protein, mitochondrial; ATP synthase membrane subunit c locus 2; ATP synthase proteolipid P2; ATP synthase proton-transporting mitochondrial F(0) complex subunit C2; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit c (subunit 9), isoform 2; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit C2 (subunit 9); ATP synthase, H+ transporting, mitochondrial Fo complex subunit C2 (subunit 9); ATP synthase, H+ transporting, mitochondrial Fo complex, subunit C2 (subunit 9); ATP5A; ATP5G2; Atp5mc2; ATPase protein 9; ATPase subunit c; mitochondrial ATP synthase, subunit C (subunit 9), isoform 2; PSEC0033
Gene Symbols ATP5MC2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ATP5G2. Recombinant protein control fragment (Product #RP-101288).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 517
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.