Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP5G2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109431
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109431 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109431 Supplier Invitrogen™ Supplier No. PA5109431
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ATAPHPLKMF ACSKFVSTPS LVKSTSQLLS RPLSAVVLKR PEILTDESLS SLAVSCPLTS LVSSRSFQTS AISRDIDTAA KFIGAGAATV GVAGSGAGIG T.

ATP5MC2 encodes the enzyme ATP5MC2, which is a subunit of the mitochondrial ATP synthase complex (Complex V). The ATP synthase complex is pivotal in the process of oxidative phosphorylation and ADP to ATP conversion, which is essential for energy production in the cell. The ATP5MC2 gene is located on chromosome 12. This subunit helps in forming the membrane channel of the F0 portion of the ATP synthase complex, facilitating proton translocation across the mitochondrial membrane. Mutations or dysfunction in this gene can lead to mitochondrial deficiencies and energy production disorders, which may have implications in various metabolic and neurodegenerative conditions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP5G2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ATP5MC2
Gene Accession No. Q06055
Gene Alias 1810041M08Rik; ATP synthase c subunit; ATP synthase F(0) complex subunit C2, mitochondrial; ATP synthase lipid-binding protein; ATP synthase lipid-binding protein, mitochondrial; ATP synthase membrane subunit c locus 2; ATP synthase proteolipid P2; ATP synthase proton-transporting mitochondrial F(0) complex subunit C2; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit c (subunit 9), isoform 2; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit C2 (subunit 9); ATP synthase, H+ transporting, mitochondrial Fo complex subunit C2 (subunit 9); ATP synthase, H+ transporting, mitochondrial Fo complex, subunit C2 (subunit 9); ATP5A; ATP5G2; Atp5mc2; ATPase protein 9; ATPase subunit c; mitochondrial ATP synthase, subunit C (subunit 9), isoform 2; PSEC0033
Gene Symbols ATP5MC2
Host Species Rabbit
Immunogen Synthetic peptide (50-150).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 517
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.