Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP5G2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501732539 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-2539 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-2539 Supplier Proteintech Group Inc Supplier No. 197851AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is specific to ATP5G2.

ATP5G2, also named as ATPase protein 9 and ATPase subunit c, belongs to the ATPase C chain family. Mitochondrial membrane ATP synthase (F1F0 ATP synthase or Complex V) produces ATP from ADP in the presence of a proton gradient across the membrane which is generated by electron transport complexes of the respiratory chain. ATP5G2 is the major protein stored in the storage bodies of animals or humans affected with ceroid lipofuscinosis (Batten disease).

Specifications

Antigen ATP5G2
Applications Immunoprecipitation
Classification Polyclonal
Concentration 0.19 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ATP5G2
Gene Accession No. P56383, Q06055
Gene Alias ATP synthase proteolipid P2, ATP5G2, ATPase protein 9, ATPase subunit c
Gene Symbols ATP5G2
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 517, 67942
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.