Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP5H Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297512IR
Description
ATP5H Polyclonal specifically detects ATP5H in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ATP5H | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
10476 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 755 | |
ATP synthase D chain, mitochondrial, ATP synthase subunit d, mitochondrial, ATP synthase, H+ transporting, mitochondrial F0 complex, subunit d, ATP synthase, H+ transporting, mitochondrial F1F0, subunit d, ATP synthase, H+ transporting, mitochondrial Fo complex, subunit d, ATP5JD, ATPase subunit d, ATPQ, My032 protein | |
Produced in rabbits immunized with E. coli-derived Human ATP5H fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction