Learn More
Invitrogen™ ATP5H Polyclonal Antibody

Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human HepG2 whole cell, human U-87MG whole cell, human A549 whole cell, monkey COS-7 whole cell, human SW620 whole cell, human Jurkat whole cell, human 293T whole cell, rat brain tissue, rat liver tissue, rat heart tissue, rat PC-12 whole cell, mouse brain tissue, mouse liver tissue, mouse heart tissue, mouse ANA-1 whole cell. IHC: human rectal cancer tissue. ICC/IF: T-47D cell.
Specifications
Specifications
| Antigen | ATP5H |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 500 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 4mg trehalose and no preservative |
| Gene | ATP5PD |
| Gene Accession No. | O75947, P31399, Q9DCX2 |
| Gene Alias | 0610009D10Rik; ATP synthase D chain, mitochondrial; ATP synthase peripheral stalk subunit d; ATP synthase subunit d, mitochondrial; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit d; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit d isoform a; ATP synthase, H+ transporting, mitochondrial F1F0, subunit d; ATP synthase, H+ transporting, mitochondrial Fo complex subunit D; ATP synthase, H+ transporting, mitochondrial Fo complex, subunit d; Atp5h; Atp5jd; Atp5pd; ATPase subunit d; ATPQ; My032; My032 protein; unnamed protein product; zgc:73071 |
| Gene Symbols | ATP5PD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.