Learn More
ATP5H Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305491A

Description
The recommended shelf life for this product is 1 year from date of receipt.
Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. It is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, which comprises the proton channel. The F1 complex consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled in a ratio of 3 alpha, 3 beta, and a single representative of the other 3. The Fo seems to have nine subunits (a, b, c, d, e, f, g, F6 and 8). This gene encodes the d subunit of the Fo complex. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene. In addition, three pseudogenes are located on chromosomes 9, 12 and 15.Specifications
ATP5H | |
Polyclonal | |
Unconjugated | |
ATP5H | |
ATPQ, ATP synthase D chain, mitochondrial, ATP synthase subunit d, mitochondrial, ATP synthase, H+ transporting, mitochondrial F0 complex, subunit d, ATP synthase, H+ transporting, mitochondrial F1F0, subunit d, ATPase subunit d, My032 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10476 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
O75947 | |
ATP5H | |
Between 100 and 150 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.