Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP5I Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116695
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116695 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116695 Supplier Invitrogen™ Supplier No. PA5116695
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: U-87MG, LO2, 293T, HeLa Immunogen sequence: MVPPVQVSPL IKLGRYSALF LGVAYGATRY NYLKPRAEEE RRIAAEEKKK QDELKRIARE LAEDDSILK.

Mitochondrial ATP synthases (ATPases) transduce the energy contained in membrane electrochemical proton gradients into the energy required for synthesis of high-energy phosphate bonds. ATPases contain two linked complexes: F1, the hydrophilic catalytic core; and F0, the membrane-embedded protein channel. The two complexes are linked by a central stalk and a peripheral stalk. During catalysis, ATP synthesis in the catalytic domain of F1 is coupled, via a rotary mechanism of the central stalk subunits, with proton translocation across the membrane. ATP5I, also known as mitochondrial ATP synthase subunit E or ATP5K, is a 69 amino acid protein member of the ATPase E subunit family. Localized to the inner membrane of the mitochondria, ATP5I is a part of the F0 complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP5I
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.49 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Atp5me
Gene Accession No. P29419, P56385, Q06185
Gene Alias 2610008D24Rik; ATP synthase e chain, mitochondrial; ATP synthase membrane subunit e; ATP synthase subunit e, mitochondrial; ATP synthase subunit e, mitochondrial, N-terminally processed; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit E; ATP synthase, H+ transporting, mitochondrial F1F0 complex, subunit e; ATP synthase, H+ transporting, mitochondrial Fo complex subunit E; ATP synthase, H+ transporting, mitochondrial Fo complex, subunit E; Atp5i; Atp5k; Atp5me; ATPase subunit e; F1F0-ATP synthase, murine e subunit; Lfm1; Lfm-1
Gene Symbols Atp5me
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-69 of human ATP5I (NP_0090311).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11958, 140608, 521
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.