Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP5L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560783
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560783 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560783 Supplier Invitrogen™ Supplier No. PA560783
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AQFVRNLVEK TPALVNAAVT YSKPRLATFW YYAKVELVPP TPAEIPRAIQ SLKKIVNSAQ TGSFK Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 72%.

UCK2 catalyzes the phosphorylation of uridine monophosphate to uridine diphosphate. This is the first step in the production of the pyrimidine nucleoside triphosphates required for RNA and DNA synthesis. In addition, an allele of this gene may play a role in mediating nonhumoral immunity to Hemophilus influenzae type B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP5L
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ATP5MG
Gene Accession No. O75964
Gene Alias 4933437C06Rik; ATP synthase g chain, mitochondrial; ATP synthase membrane subunit g; ATP synthase subunit g, mitochondrial; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit G; ATP synthase, H+ transporting, mitochondrial F1F0, subunit g; ATP synthase, H+ transporting, mitochondrial Fo complex subunit G; ATP synthase, H+ transporting, mitochondrial Fo complex, subunit G; ATP5JG; ATP5L; Atp5mg; ATPase subunit g; F1F0-ATP synthase g subunit; F1F0-type ATP synthase subunit g; F1Fo-ATP synthase complex Fo membrane domain g subunit
Gene Symbols ATP5MG
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ATP5L. Recombinant protein control fragment (Product #RP-97440).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10632
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.