Learn More
Invitrogen™ ATP5L Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121225
Description
Positive test controls include: 293T, A-549, HepG2, HeLa, Mouse brain, Mouse liver, Rat brain, Rat kidney. The target is usually found in the following locations: Mitochondrion, Mitochondrion inner membrane. Immunogen sequence: MAQFVRNLVE KTPALVNAAV TYSKPRLATF WYYAKVELVP PTPAEIPRAI QSLKKIVNSA QTGSFKQLTV KEAVLNGLVA TEVLMWFYVG EIIGKRGIIG YDV.
UCK2 catalyzes the phosphorylation of uridine monophosphate to uridine diphosphate. This is the first step in the production of the pyrimidine nucleoside triphosphates required for RNA and DNA synthesis. In addition, an allele of this gene may play a role in mediating nonhumoral immunity to Hemophilus influenzae type B.
Specifications
ATP5L | |
Polyclonal | |
Unconjugated | |
ATP5MG | |
4933437C06Rik; ATP synthase g chain, mitochondrial; ATP synthase membrane subunit g; ATP synthase subunit g, mitochondrial; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit G; ATP synthase, H+ transporting, mitochondrial F1F0, subunit g; ATP synthase, H+ transporting, mitochondrial Fo complex subunit G; ATP synthase, H+ transporting, mitochondrial Fo complex, subunit G; ATP5JG; ATP5L; Atp5mg; ATPase subunit g; F1F0-ATP synthase g subunit; F1F0-type ATP synthase subunit g; F1Fo-ATP synthase complex Fo membrane domain g subunit | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10632, 27425, 300677 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.2 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
O75964, Q6PDU7, Q9CPQ8 | |
ATP5MG | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-103 of human ATP5L (NP_006467.4). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.