Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP5L2 Polyclonal Antibody
ATP5L2 Polyclonal Antibody
Supplier: Thermo Scientific PA575985
Description
ATP5L2 Polyclonal Antibody for Western Blot

Specifications
ATP5L2 | |
Polyclonal | |
Unconjugated | |
ATP5L2 | |
ATP synthase membrane subunit g-like protein; ATP synthase subunit g 2, mitochondrial; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit G2 pseudogene; ATP synthase, H+ transporting, mitochondrial F1F0, subunit g; ATP synthase, H+ transporting, mitochondrial Fo complex subunit G2; ATP synthase, H+ transporting, mitochondrial Fo complex, subunit G2; ATP5K2; ATP5L2; ATP5MGL; ATPase subunit g 2; Putative ATP synthase subunit g 2, mitochondrial | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
267020 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z4Y8 | |
ATP5L2 | |
Synthetic peptide corresponding to amino acids 56-100 of Human ATP5L2. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction