Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP5L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101442
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101442 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101442 Supplier Invitrogen™ Supplier No. PA5101442
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ATP5L2.

Mitochondrial membrane ATP synthase (F(1)F(0) ATP synthase or Complex V) produces ATP from ADP in the presence of a proton gradient across the membrane which is generated by electron transport complexes of the respiratory chain. F-type ATPases consist of two structural domains, F(1) - containing the extramembraneous catalytic core, and F(0) - containing the membrane proton channel, linked together by a central stalk and a peripheral stalk. During catalysis, ATP synthesis in the catalytic domain of F(1) is coupled via a rotary mechanism of the central stalk subunits to proton translocation. Part of the complex F(0) domain. Minor subunit located with subunit a in the membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP5L2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ATP5L2
Gene Accession No. Q7Z4Y8
Gene Alias ATP synthase membrane subunit g-like protein; ATP synthase subunit g 2, mitochondrial; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit G2 pseudogene; ATP synthase, H+ transporting, mitochondrial F1F0, subunit g; ATP synthase, H+ transporting, mitochondrial Fo complex subunit G2; ATP synthase, H+ transporting, mitochondrial Fo complex, subunit G2; ATP5K2; ATP5L2; ATP5MGL; ATPase subunit g 2; Putative ATP synthase subunit g 2, mitochondrial
Gene Symbols ATP5L2
Host Species Rabbit
Immunogen A synthesized peptide derived from human ATP5L2(Accession Q7Z4Y8), corresponding to amino acid residues S64-G100.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 267020
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.