Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP5O Mouse anti-Human, Clone: 1D3B8, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | ATP5O |
---|---|
Clone | 1D3B8 |
Concentration | 0.75 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-7112
![]() ![]() |
Proteintech Group Inc
666961IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The protein encoded by this gene is a component of the F-type ATPase found in the mitochondrial matrix. F-type ATPases are composed of a catalytic core and a membrane proton channel. The encoded protein appears to be part of the connector linking these two components and may be involved in transmission of conformational changes or proton conductance.Specifications
ATP5O | |
0.75 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ATP5O, ATPO, OSCP | |
ATP5O | |
IgG1 | |
Protein G | |
Antibody |
1D3B8 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P48047 | |
539 | |
ATP5O Fusion Protein Ag1458 | |
Primary | |
-20°C | |
ATP5O |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title