Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP5O Monoclonal Antibody (OTI2E9), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA804572

Description
The protein encoded by this gene is a component of the F-type ATPase found in the mitochondrial matrix. F-type ATPases are composed of a catalytic core and a membrane proton channel. The encoded protein appears to be part of the connector linking these two components and may be involved in transmission of conformational changes or proton conductance.Specifications
ATP5O | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P48047 | |
ATP5O | |
Human recombit protein fragment corresponding to amino acids 24-213 of human ATP5O produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2E9 | |
Unconjugated | |
ATP5O | |
ATP5O, ATPO, HMC08D05, OSCP | |
Mouse | |
Affinity Chromatography | |
RUO | |
539 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction