Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ATP5S Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA536926
Description
This antibody detects endogenous protein at a molecular weight of 25 kDa. Purity is >95% by SDS-PAGE.
ATP5S, also known as ATP synthase subunits, mitochondrial, ATP synthase-coupling factor B or ATP synthase, H+ transporting, mitochondrial F0 complex, subunits (factor B), is a 215 amino acid mitochondrial inner membrane protein that belongs to the ATP synthase subunits family. Involved in regulation of mitochondrial membrane ATP synthase, ATP5S is necessary for H+ conduction of ATP synthase. The ATP5S gene encodes subunits, also known as factor B, of the proton channel. This subunit is necessary for energy transduction in ATP synthase complexes. The ATP5S gene is conserved in chimpanzee, canine, bovine, mouse, rat, chicken, zebrafish and mosquito, and maps to human chromosome 14q21.3.
Specifications
| ATP5S | |
| Polyclonal | |
| Unconjugated | |
| DMAC2L | |
| 1110015E18Rik; ATP synthase coupling factor B, mitochondrial; ATP synthase coupling factor B-like 1; ATP synthase subunit s, mitochondrial; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit S; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit s (factor B); ATP synthase, H+ transporting, mitochondrial Fo complex subunit s (factor B); ATP synthase, H+ transporting, mitochondrial Fo complex, subunit s (factor B); ATP synthase-coupling factor B; ATP5S; Atpw; Distal membrane arm assembly complex 2-like protein; DMAC2L; Factor B; facyor B; FB; HSU79253; Mitochondrial ATP synthase regulatory component factor B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27109, 362749, 68055 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5XIM4, Q99766, Q9CRA7 | |
| DMAC2L | |
| Synthetic peptide corresponding to amino acids 73-118 of human ATP5S. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction