Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP5SL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546193
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546193 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546193 Supplier Invitrogen™ Supplier No. PA546193
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KRTILQFLTN YFYDVEALRD YLLQREMYKV HEKNRSYTWL EKQHGPYGAG Sequence homology: Human: 100%.

Required for the assembly of the mitochondrial NADH:ubiquinone oxidoreductase complex (complex I). Involved in the assembly of the distal region of complex I. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP5SL
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Dmac2
Gene Accession No. Q9NW81
Gene Alias ATP synthase subunit s-like protein; ATP5S like; Atp5sl; ATP5S-like; distal membrane arm assembly complex 2; distal membrane-arm assembly complex protein 2; DMAC2
Gene Symbols Dmac2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ATP5SL (aa 38-87).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55101
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.