Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP6AP1L Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP23089925UL
Description
ATP6AP1L Polyclonal specifically detects ATP6AP1L in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
ATP6AP1L | |
Polyclonal | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Q52LC2 | |
ATP6AP1L | |
This antibody was developed against a recombinant protein corresponding to amino acids: AVFNPTGVYAPSGYSYRCQRVGSLQQDQALLLPSDTDDGSSLWEVTFIDFQIQGFAIKGGRFTKAQDCASSFSPA | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
ATPase, H+ transporting, lysosomal accessory protein 1-like, MGC138396, vacuolar proton pump subunit S1-like protein, V-type proton ATPase subunit S1-like protein | |
Rabbit | |
Affinity Purified | |
RUO | |
92270 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction