Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP6AP1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569341
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569341 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569341 Supplier Invitrogen™ Supplier No. PA569341
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Dog: 93%; Horse: 92%; Human: 100%; Pig: 79%.

ATP6AP1L gene ontology annotations related to this gene include ATP hydrolysis coupled proton transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP6AP1L
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ATP6AP1L
Gene Accession No. Q52LC2
Gene Alias ATP6AP1L; ATPase H+ transporting accessory protein 1 like; ATPase, H+ transporting, lysosomal accessory protein 1-like; Vacuolar proton pump subunit S1-like protein; V-type proton ATPase subunit S1-like protein
Gene Symbols ATP6AP1L
Host Species Rabbit
Immunogen synthetic peptide directed towards the C terminal region of human ATP6AP1L.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92270
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.