Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP6IP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115835
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115835 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115835 Supplier Invitrogen™ Supplier No. PA5115835
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Renin receptor.

This gene encodes a protein that is associated with adenosine triphosphatases (ATPases). Proton-translocating ATPases have fundamental roles in energy conservation, secondary active transport, acidification of intracellular compartments, and cellular pH homeostasis. There are three classes of ATPases- F, P, and V. The vacuolar (V-type) ATPases have a transmembrane proton-conducting sector and an extramembrane catalytic sector. The encoded protein has been found associated with the transmembrane sector of the V-type ATPases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP6IP2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Atp6ap2
Gene Accession No. O75787, Q6AXS4, Q9CYN9
Gene Alias (P)RR; (pro)renin receptor; 5730403E06Rik; APT6M8-9; ATP6AP2; Atp6ip2; ATP6M8-9; ATPase H(+)-transporting lysosomal accessory protein 2; ATPase H(+)-transporting lysosomal-interacting protein 2; ATPase H+ transporting accessory protein 2; ATPase, H(+)-transporting, lysosomal-interacting protein 2; ATPase, H+ transporting, lysosomal (vacuolar proton pump) membrane sector associated protein M8-9; ATPase, H+ transporting, lysosomal accessory protein 2; ATPase, H+ transporting, lysosomal interacting protein 1; ATPase, H+ transporting, lysosomal interacting protein 2; CAPER; ELDF10; embryonic liver differentiation factor 10; ER-localized type I transmembrane adaptor; HT028; M8-9; MGC99577; MRXE; MRXSH; MSTP009; N14F; prorenin receptor; PRR; PSEC0072; Renin receptor; Renin receptor C-terminal fragment; Renin receptor cytoplasmic fragment; Renin receptor extracellular fragment; Renin receptor N-terminal fragment; Renin/prorenin receptor; RENR; vacuolar ATP synthase membrane sector-associated protein M8-9; vacuolar proton ATP synthase membrane sector associated protein M8-9; V-ATPase M8.9 subunit; XMRE; XPDS
Gene Symbols Atp6ap2
Host Species Rabbit
Immunogen A synthesized peptide derived from human ATP6AP2(Accession O75787), corresponding to amino acid residues V179-D229.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10159, 302526, 70495
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.