Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP6IP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518205
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518205 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518205 Supplier Invitrogen™ Supplier No. PA518205
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.

This gene encodes a protein that is associated with adenosine triphosphatases (ATPases). Proton-translocating ATPases have fundamental roles in energy conservation, secondary active transport, acidification of intracellular compartments, and cellular pH homeostasis. There are three classes of ATPases- F, P, and V. The vacuolar (V-type) ATPases have a transmembrane proton-conducting sector and an extramembrane catalytic sector. The encoded protein has been found associated with the transmembrane sector of the V-type ATPases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP6IP2
Applications Flow Cytometry, Western Blot, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Atp6ap2
Gene Accession No. O75787, Q6AXS4
Gene Alias (P)RR; (pro)renin receptor; 5730403E06Rik; APT6M8-9; ATP6AP2; Atp6ip2; ATP6M8-9; ATPase H(+)-transporting lysosomal accessory protein 2; ATPase H(+)-transporting lysosomal-interacting protein 2; ATPase H+ transporting accessory protein 2; ATPase, H(+)-transporting, lysosomal-interacting protein 2; ATPase, H+ transporting, lysosomal (vacuolar proton pump) membrane sector associated protein M8-9; ATPase, H+ transporting, lysosomal accessory protein 2; ATPase, H+ transporting, lysosomal interacting protein 1; ATPase, H+ transporting, lysosomal interacting protein 2; CAPER; ELDF10; embryonic liver differentiation factor 10; ER-localized type I transmembrane adaptor; HT028; M8-9; MGC99577; MRXE; MRXSH; MSTP009; N14F; prorenin receptor; PRR; PSEC0072; Renin receptor; Renin receptor C-terminal fragment; Renin receptor cytoplasmic fragment; Renin receptor extracellular fragment; Renin receptor N-terminal fragment; Renin/prorenin receptor; RENR; vacuolar ATP synthase membrane sector-associated protein M8-9; vacuolar proton ATP synthase membrane sector associated protein M8-9; V-ATPase M8.9 subunit; XMRE; XPDS
Gene Symbols Atp6ap2
Host Species Goat
Immunogen Synthetic peptide sequence (SIIYRMTNQKIRMD) corresponding to the C-terminus amino acids of ATP6AP2 (aa 337-350).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10159, 302526
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.