Learn More
Invitrogen™ ATP6IP2 Polyclonal Antibody

Description
Recommended positive controls: The peptide used to generate this antibody is available for purchase (GTX25959-PEP)..
Specifications
Specifications
| Antigen | ATP6IP2 |
| Applications | Flow Cytometry, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | Atp6ap2 |
| Gene Accession No. | O75787, Q6AXS4, Q9CYN9 |
| Gene Alias | (P)RR; (pro)renin receptor; 5730403E06Rik; APT6M8-9; ATP6AP2; Atp6ip2; ATP6M8-9; ATPase H(+)-transporting lysosomal accessory protein 2; ATPase H(+)-transporting lysosomal-interacting protein 2; ATPase H+ transporting accessory protein 2; ATPase, H(+)-transporting, lysosomal-interacting protein 2; ATPase, H+ transporting, lysosomal (vacuolar proton pump) membrane sector associated protein M8-9; ATPase, H+ transporting, lysosomal accessory protein 2; ATPase, H+ transporting, lysosomal interacting protein 1; ATPase, H+ transporting, lysosomal interacting protein 2; CAPER; ELDF10; embryonic liver differentiation factor 10; ER-localized type I transmembrane adaptor; HT028; M8-9; MGC99577; MRXE; MRXSH; MSTP009; N14F; prorenin receptor; PRR; PSEC0072; Renin receptor; Renin receptor C-terminal fragment; Renin receptor cytoplasmic fragment; Renin receptor extracellular fragment; Renin receptor N-terminal fragment; Renin/prorenin receptor; RENR; vacuolar ATP synthase membrane sector-associated protein M8-9; vacuolar proton ATP synthase membrane sector associated protein M8-9; V-ATPase M8.9 subunit; XMRE; XPDS |
| Gene Symbols | Atp6ap2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.