Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP6V0A2 Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299457FR
Description
ATP6V0A2 Polyclonal specifically detects ATP6V0A2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ATP6V0A2 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
A2, ARCL, ATP6a2, ATP6N1D, ATP6V0, ATPase, H+ transporting, lysosomal V0 subunit a isoform 2, ATPase, H+ transporting, lysosomal V0 subunit a2, infantile malignant osteopetrosis, J6B7, Lysosomal H(+)-transporting ATPase V0 subunit a2, regeneration and tolerance factor, RTF, STV1, TJ6A2V-ATPase, TJ6M, TJ6S, vacuolar proton translocating ATPase 116 kDa subunit a, Vacuolar proton translocating ATPase 116 kDa subunit a isoform 2, v-ATPase 116 kDa, V-ATPase 116 kDa isoform a2, Vph1, v-type proton ATPase 116 kDa subunit a, V-type proton ATPase 116 kDa subunit a isoform 2, WSS | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human ATP6V0A2. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
23545 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction