Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP6V0A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529271
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529271 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529271 Supplier Invitrogen™ Supplier No. PA529271
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: unboiled DDDDK-tagged ATP6V0A-transfested 293T. Predicted reactivity: Mouse (93%), Rat (94%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The multisubunit vacuolar-type proton pump (H(+)-ATPase or V-ATPase) is essential for acidification of diverse cellular components, including endosomes, lysosomes, clathrin-coated vesicles, secretory vesicles, and chromaffin granules, and it is found at high density in the plasma membrane of certain specialized cells. H(+)-ATPases are comprised of a peripheral V(1) domain and an integral membrane V(0) domain; ATP6V0A2 is a component of the V(0) domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP6V0A2
Applications Western Blot
Classification Polyclonal
Concentration 0.89 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene ATP6V0A2
Gene Accession No. Q9Y487
Gene Alias 8430408C20Rik; A2; A2V-ATPase; AI385560; ARCL; ARCL2A; ATP6 ATP6N1D; ATP6A2; Atp6n1b; Atp6n1d; Atp6n2; ATP6V0A2; ATPase H+ transporting V0 subunit a2; ATPase, H+ transporting, lysosomal (vacuolar proton pump) non-catalytic accessory protein 2 (38kD); ATPase, H+ transporting, lysosomal V0 subunit a; ATPase, H+ transporting, lysosomal V0 subunit a2; AW489264; C76904; Cc1-3; Immune suppressor factor J6B7; ISF; J6B7; lysosomal H(+)-transporting ATPase V0 subunit a2; regeneration and tolerance factor; RTF; SHIF; Stv1; T-cell expressing clone j6; Tj6; TJ6M; TJ6s; vacuolar proton translocating ATPase 116 kDa subunit a; Vacuolar proton translocating ATPase 116 kDa subunit a isoform 2; vacuolar proton translocating ATPase 116-kDa subunit a2; vacuolar proton translocating ATPase 116-kDa subunit a2 isoform; V-ATPase 116 kDa; V-ATPase 116 kDa isoform a2; V-ATPase 116 kDa subunit a2; V-ATPase a2; VPH1; v-type proton ATPase 116 kDa subunit a; V-type proton ATPase 116 kDa subunit a isoform 2; V-type proton ATPase 116 kDa subunit a isoform 2; V-type proton ATPase 116 kDa subunit a; V-type proton ATPase 116 kDa subunit a2; WSS
Gene Symbols ATP6V0A2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ATP6V0A2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23545
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.