Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ATP6V0A2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542156
Description
Peptide sequence: INRADIPLPE GEASPPAPPL KQVLEMQEQL QKLEVELREV TKNKEKLRKN Sequence homology: Cow: 86%; Dog: 93%; Guinea Pig: 83%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 92%; Rat: 100%; Yeast: 79%; Zebrafish: 77%.
The multisubunit vacuolar-type proton pump (H(+)-ATPase or V-ATPase) is essential for acidification of diverse cellular components, including endosomes, lysosomes, clathrin-coated vesicles, secretory vesicles, and chromaffin granules, and it is found at high density in the plasma membrane of certain specialized cells. H(+)-ATPases are comprised of a peripheral V(1) domain and an integral membrane V(0) domain; ATP6V0A2 is a component of the V(0) domain.
Specifications
| ATP6V0A2 | |
| Polyclonal | |
| Unconjugated | |
| ATP6V0A2 | |
| 8430408C20Rik; A2; A2V-ATPase; AI385560; ARCL; ARCL2A; ATP6 ATP6N1D; ATP6A2; Atp6n1b; Atp6n1d; Atp6n2; ATP6V0A2; ATPase H+ transporting V0 subunit a2; ATPase, H+ transporting, lysosomal (vacuolar proton pump) non-catalytic accessory protein 2 (38kD); ATPase, H+ transporting, lysosomal V0 subunit a; ATPase, H+ transporting, lysosomal V0 subunit a2; AW489264; C76904; Cc1-3; Immune suppressor factor J6B7; ISF; J6B7; lysosomal H(+)-transporting ATPase V0 subunit a2; regeneration and tolerance factor; RTF; SHIF; Stv1; T-cell expressing clone j6; Tj6; TJ6M; TJ6s; vacuolar proton translocating ATPase 116 kDa subunit a; Vacuolar proton translocating ATPase 116 kDa subunit a isoform 2; vacuolar proton translocating ATPase 116-kDa subunit a2; vacuolar proton translocating ATPase 116-kDa subunit a2 isoform; V-ATPase 116 kDa; V-ATPase 116 kDa isoform a2; V-ATPase 116 kDa subunit a2; V-ATPase a2; VPH1; v-type proton ATPase 116 kDa subunit a; V-type proton ATPase 116 kDa subunit a isoform 2; V-type proton ATPase 116 kDa subunit a isoform 2; V-type proton ATPase 116 kDa subunit a; V-type proton ATPase 116 kDa subunit a2; WSS | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 23545 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9Y487 | |
| ATP6V0A2 | |
| Synthetic peptide directed towards the N-terminal of human ATP6V0A2 (aa 72-121). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction