Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP6V0A4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596730
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596730 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596730 Supplier Invitrogen™ Supplier No. PA596730
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MVSVFRSEEM CLSQLFLQVE AAYCCVAELG ELGLVQFKDL NMNVNSFQRK FVNEVRRCES LERILRFLED EMQNEIVVQL LEKSPLTPLP REMITLETVL EKLEGELQEA NQNQQALKQS FLELTELKYL LKKTQDFFET ETNLADDFFT EDTSGLLELK AVPAYMTGKL GFIAGVINRE RMASFERLLW RICRGNVYLK FSEMDAPLED PVTKEEIQKN IFIIFYQGEQ LRQKIKKICD GFRATVYPCP EPAVERREML ESVNVRLEDL ITVITQTESH; Positive Samples: A-375, Mouse brain; Cellular Location: Apical cell membrane, Multi-pass membrane protein.

This gene encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of intracellular compartments of eukaryotic cells. V-ATPase dependent acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c', and d. This gene is one of four genes in man and mouse that encode different isoforms of the a subunit. Alternatively spliced transcript variants encoding the same protein have been described. Mutations in this gene are associated with renal tubular acidosis associated with preserved hearing. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP6V0A4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.16 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ATP6V0A4
Gene Accession No. Q920R6, Q9HBG4
Gene Alias a4; Atp6n1b; ATP6N2; Atp6v0a4; ATPase H+ transporting V0 subunit a4; ATPase, H+ transporting, lysosomal (vacuolar proton pump) noncatalytic accessory protein 1B; ATPase, H+ transporting, lysosomal (vacuolar proton pump) non-catalytic accessory protein 1B; ATPase, H+ transporting, lysosomal (vacuolar proton pump) non-catalytic accessory protein 2 (38kD); ATPase, H+ transporting, lysosomal V0 subunit A4; H(+)-transporting two-sector ATPase, noncatalytic accessory protein 1B; RDRTA2; RTA1C; RTADR; STV1; tcag7.346; vacuol; vacuolar proton pump 116 kDa accessory subunit; vacuolar proton pump, subunit 2; vacuolar proton translocating ATPase 100 kDa a4 subunit; Vacuolar proton translocating ATPase 116 kDa subunit a isoform 4; Vacuolar proton translocating ATPase 116 kDa subunit a kidney isoform; V-ATPase 116 kDa; V-ATPase 116 kDa isoform a4; V-ATPase alpha 4; VPH1; VPP2; V-type proton ATPase 116 kDa subunit a; V-type proton ATPase 116 kDa subunit a isoform 4
Gene Symbols ATP6V0A4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human ATP6V0A4 (NP_0656832).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140494, 50617
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.