Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP6V0A4 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA522158

Catalog No. PIPA522158


Only null left
Add to Cart

Description

Description

Recommended positive controls: A431, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (87%), Bovine (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of intracellular compartments of eukaryotic cells. V-ATPase dependent acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c', c'', and d. This gene is one of four genes in man and mouse that encode different isoforms of the a subunit. Alternatively spliced transcript variants encoding the same protein have been described. Mutations in this gene are associated with renal tubular acidosis associated with preserved hearing. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ATP6V0A4
Polyclonal
Unconjugated
ATP6V0A4
a4; Atp6n1b; ATP6N2; Atp6v0a4; ATPase H+ transporting V0 subunit a4; ATPase, H+ transporting, lysosomal (vacuolar proton pump) noncatalytic accessory protein 1B; ATPase, H+ transporting, lysosomal (vacuolar proton pump) non-catalytic accessory protein 1B; ATPase, H+ transporting, lysosomal (vacuolar proton pump) non-catalytic accessory protein 2 (38kD); ATPase, H+ transporting, lysosomal V0 subunit A4; H(+)-transporting two-sector ATPase, noncatalytic accessory protein 1B; RDRTA2; RTA1C; RTADR; STV1; tcag7.346; vacuol; vacuolar proton pump 116 kDa accessory subunit; vacuolar proton pump, subunit 2; vacuolar proton translocating ATPase 100 kDa a4 subunit; Vacuolar proton translocating ATPase 116 kDa subunit a isoform 4; Vacuolar proton translocating ATPase 116 kDa subunit a kidney isoform; V-ATPase 116 kDa; V-ATPase 116 kDa isoform a4; V-ATPase alpha 4; VPH1; VPP2; V-type proton ATPase 116 kDa subunit a; V-type proton ATPase 116 kDa subunit a isoform 4
Rabbit
Antigen affinity chromatography
RUO
50617
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q9HBG4
ATP6V0A4
Recombinant protein encompassing a sequence within the N-terminus region of human ATP6V0A4. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.