Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP6V0C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA523972
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA523972 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA523972 Supplier Invitrogen™ Supplier No. PA523972
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with bovine, canine, equine and rat based on sequence homology.

ATP6V0C is a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c', c', and d. ATP6V0C encodes the V0 subunit c.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP6V0C
Applications Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ATP6V0C
Gene Accession No. P27449, P63082
Gene Alias Atp6c; Atp6c2; Atp6l; ATP6V0C; ATPase H+ transporting V0 subunit c; ATPase, H+ transporting, lysosomal (vacuolar proton pump) 16 kDa; ATPase, H+ transporting, lysosomal (vacuolar proton pump), subunit C; ATPase, H+ transporting, lysosomal 1; ATPase, H+ transporting, lysosomal 16kD, V0 subunit C; ATPase, H+ transporting, lysosomal 16kDa, V0 subunit c; ATPase, H+ transporting, lysosomal 2; ATPase, H+ transporting, lysosomal V0 subunit C; ATPase, H+ transporting, V0 subunit C; ATPL; Atpl-rs1; H(+)-ATPase (mvp); H(+)-transporting two-sector ATPase, 16 kDa subunit; Mvp; PL16; vacuolar ATP synthase 16 kDa proteolipid subunit; vacuolar H+ ATPase proton channel subunit; vacuolar proton pump 16 kDa proteolipid subunit; VATL; V-ATPase 16 kDa proteolipid subunit; Vma3; VPPC; V-type proton ATPase 16 kDa proteolipid subunit
Gene Symbols ATP6V0C
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 100-126 amino acids from the C-terminal region of human ATP6V0C.
Purification Method Protein A, Antigen affinity chromatography
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11984, 527
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.