Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP6V0D1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP215517

Catalog No. NBP215517


Only null left
Add to Cart

Description

Description

ATP6V0D1 Polyclonal specifically detects ATP6V0D1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

ATP6V0D1
Polyclonal
Unconjugated
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
32 kDa accessory protein, ATP6D, ATP6DV, ATP6V0, ATPase, H+ transporting, lysosomal (vacuolar proton pump), member D, ATPase, H+ transporting, lysosomal 38kDa, V0 subunit d isoform 1, ATPase, H+ transporting, lysosomal 38kDa, V0 subunit d1, FLJ43534, H(+)-transporting two-sector ATPase, subunit D, P39, Vacuolar proton pump subunit d 1, V-ATPase 40 kDa accessory protein, V-ATPase AC39 subunit, V-ATPase, subunit D, VATX, Vma6, VPATPDV-ATPase subunit d 1, V-type proton ATPase subunit d 1
Rabbit
40 kDa
0.1 mL
Primary
Human. Expected to cross react based on sequence identity: Mouse (100%), Rat (100%), Bovine (100%).
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Western Blot
1mg/mL
Western Blot 1:500-1:3000
P61421
ATP6V0D1
Recombinant protein encompassing a sequence within the center region of human ATP6V0D1. The exact sequence is proprietary.
Affinity Purified
RUO
9114
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only