Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP6V0D2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107075
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107075 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107075 Supplier Thermo Scientific Supplier No. PA5107075
Add to Cart
Add to Cart

ATP6V0D2 Polyclonal Antibody

ATP6V0D2 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP6V0D2
Applications Western Blot, Surface Plasmon Resonance
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene ATP6V0D2
Gene Accession No. Q8N8Y2
Gene Alias 1620401A02Rik; AI324824; ATP6D2; Atp6v0d2; ATPase H+ transporting V0 subunit d2; ATPase, H+ transporting, lysosomal 38kDa, V0 subunit d2; ATPase, H+ transporting, lysosomal V0 subunit D2; ATPase, H+ transporting, V0 subunit D, isoform 2; FLJ38708; osteoclast-specific vacuolar ATP synthase; similar to ATPase, H+ transporting, V0 subunit D isoform 2; vacuolar proton pump subunit d 2; vacuolar proton-translocating ATPase d subunit d2; V-ATPase; V-ATPase subunit d 2; VMA6; V-type proton ATPase subunit d 2
Gene Symbols ATP6V0D2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human ATP6V0D2 (aa 300-350).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 245972
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.