Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP6V0E2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570863
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570863 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570863 Supplier Invitrogen™ Supplier No. PA570863
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

Multisubunit vacuolar-type proton pumps, or H(+)-ATPases, acidify various intracellular compartments, such as vacuoles, clathrin-coated and synaptic vesicles, endosomes, lysosomes, and chromaffin granules. H(+)-ATPases are also found in plasma membranes of specialized cells, where they play roles in urinary acidification, bone resorption, and sperm maturation. Multiple subunits form H(+)-ATPases, with proteins of the V1 class hydrolyzing ATP for energy to transport H+, and proteins of the V0 class forming an integral membrane domain through which H+ is transported. ATP6V0E2 encodes an isoform of the H(+)-ATPase V0 e subunit, an essential proton pump component.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP6V0E2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ATP6V0E2
Gene Accession No. Q8NHE4
Gene Alias 0610006O14Rik; ATP6V0E2; ATP6V0E2L; ATPase H+ transporting V0 subunit e2; ATPase, H+ transporting V0 subunit e2; ATPase, H+ transporting, lysosomal V0 subunit E2; ATPase, H+ transporting, lysosomal, V0 subunit E; ATPase, H+ transporting, V0 subunit E; C7orf32; H+-ATPase e2 subunit; lysosomal 9 kDa H(+)-transporting ATPase V0 subunit e2; NM9.2; similar to ATPase, H+ transporting, V0 subunit; vacuolar proton pump subunit e 2; vacuolar proton-ATPase subunit; V-ATPase subunit e 2; V-type proton ATPase subunit e 2
Gene Symbols ATP6V0E2
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human ATP6V0E2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 155066
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.