Learn More
Invitrogen™ ATP6V0E2 Polyclonal Antibody

Description
This target displays homology in the following species: Human: 100% This antibody recognizes isoforms 2 and 3 of human vacuolar ATPase.
Specifications
Specifications
| Antigen | ATP6V0E2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | ATP6V0E2 |
| Gene Accession No. | Q8NHE4 |
| Gene Alias | 0610006O14Rik; ATP6V0E2; ATP6V0E2L; ATPase H+ transporting V0 subunit e2; ATPase, H+ transporting V0 subunit e2; ATPase, H+ transporting, lysosomal V0 subunit E2; ATPase, H+ transporting, lysosomal, V0 subunit E; ATPase, H+ transporting, V0 subunit E; C7orf32; H+-ATPase e2 subunit; lysosomal 9 kDa H(+)-transporting ATPase V0 subunit e2; NM9.2; similar to ATPase, H+ transporting, V0 subunit; vacuolar proton pump subunit e 2; vacuolar proton-ATPase subunit; V-ATPase subunit e 2; V-type proton ATPase subunit e 2 |
| Gene Symbols | ATP6V0E2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.