Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP6V1B Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS12549R

Description
ATP6V1B2 encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A, three B, and two G subunits, as well as a C, D, E, F, and H subunit. The V1 domain contains the ATP catalytic site. The protein encoded by this gene is one of two V1 domain B subunit isoforms and is the only B isoform highly expressed in osteoclasts.Specifications
ATP6V1B | |
Polyclonal | |
Unconjugated | |
Atp6v1b2 | |
AI194269; AI790362; ATP6B1B2; Atp6b2; Atp6v1b2; atp6v1bb; atp6v1bb protein; ATPase H+ transporting V1 subunit B2; ATPase, H transporting, lysosomal V1 subunit B2; ATPase, H+ transporting, lysosomal (vacuolar proton pump), beta 56/58 kDa, isoform 2; ATPase, H+ transporting, lysosomal 56/58kD, V1 subunit B, isoform 2; ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B1; ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B2; ATPase, H+ transporting, lysosomal V1 subunit B2; ATPase, H+ transporting, lysosomal, V1 subunit B, member b; ATPase, H+ transporting, lysosomal, V1 subunit B2; ATPase, H+ transporting, V1 subunit B, isoform 2; BOS_9033; DOOD; Endomembrane proton pump 58 kDa subunit; fj51e01; H+ transporting two-sector ATPase; H+-ATPase B subunit; H+-ATPase non-catalytic subunit B; HO57; LOC100341367; lysosomal 56/58kDa; R74844; testicular secretory protein Li 65; vacuolar H+-ATPase; vacuolar H+-ATPase 56,000 subunit; vacuolar H+ATPase B2; Vacuolar proton pump subunit B 2; vacuolar-type H+ transporting ATPase subunit B2; Vat2; Vatb; vatB2; V-ATPase B2 subunit; V-ATPase subunit B 2; Vma2; VPP3; V-type proton ATPase subunit B, brain isoform; wu:fj51e01; zgc:109771; ZLS2 | |
Rabbit | |
Protein A | |
RUO | |
526 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P21281 | |
Atp6v1b2 | |
KLH conjugated synthetic peptide derived from human ATP6V1B2/V-ATPase B2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction